Search
Search

fcgr1| Gene Symbol: fcgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114208.1 | NP_001107680.1 | 6 - 7 | 1228 | 88 | |
| GenBank mRNA | BC088875.1 | - | 6 - 7 | 1200 | 88 | |
| BC154883.1 | - | 6 - 7 | 1228 | 88 | ||
Set Membership: |
|