Search
Search

unkl| Gene Symbol: unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123443.1 | NP_001116915.1 | 10 - 11 | 1492 | 129 | |
| XM_012970032.2 | XP_012825486.1 | 11 - 12 | 3099 | 129 | ||
| GenBank mRNA | BC160454.1 | - | 10 - 11 | 1492 | 129 | |
Set Membership: |
|