Search
Search

ptpn6| Gene Symbol: ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123456.1 | NP_001116928.1 | 13 - 14 | 1798 | 91 | |
| GenBank mRNA | BC160526.1 | - | 13 - 14 | 1798 | 91 | |
Set Membership: |
|