Search
Search

ctnnd1| Gene Symbol: ctnnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126073.1 | NP_001119545.1 | 15 - 16 | 2411 | 68 | |
| XM_012960396.2 | XP_012815850.1 | 14 - 15 | 2473 | 68 | ||
| XM_012960397.2 | XP_012815851.1 | 14 - 15 | 2493 | 68 | ||
| XM_012960399.2 | XP_012815853.1 | 14 - 15 | 2472 | 68 | ||
| XM_012960400.2 | XP_012815854.1 | 13 - 14 | 2250 | 68 | ||
| XM_012960401.2 | XP_012815855.1 | 13 - 14 | 2209 | 68 | ||
| XM_012960402.2 | XP_012815856.1 | 13 - 14 | 2283 | 68 | ||
| XM_012960403.2 | XP_012815857.1 | 13 - 14 | 2262 | 68 | ||
| XM_018095212.1 | XP_017950701.1 | 13 - 14 | 2335 | 68 | ||
| XM_018095213.1 | XP_017950702.1 | 13 - 14 | 2335 | 68 | ||
| GenBank mRNA | BC166230.1 | - | 15 - 16 | 2411 | 68 | |
| CT027913.2 | - | 14 - 15 | 2360 | 68 | ||
Set Membership: |
|