Search
Search

grasp| Gene Symbol: grasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126882.1 | NP_001120354.1 | 1 - 2 | 226 | 66 | |
| GenBank mRNA | BC160992.1 | - | 1 - 2 | 226 | 66 | |
| CU075797.1 | - | 1 - 2 | 215 | 66 | ||
Set Membership: |
|