Search
Search

ythdf3| Gene Symbol: ythdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012964301.2 | XP_012819755.1 | 3 - 4 | 153 | 90 | |
| XM_012964302.2 | XP_012819756.1 | 3 - 4 | 223 | 90 | ||
| GenBank mRNA | BC161192.1 | - | 2 - 3 | 433 | 90 | |
| CT025387.2 | - | 2 - 3 | 443 | 90 | ||
Set Membership: |
|