Search
Search

gopc| Gene Symbol: gopc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126685.1 | NP_001120157.1 | 3 - 4 | 652 | 73 | |
| XM_012962715.2 | XP_012818169.1 | 4 - 5 | 737 | 73 | ||
| XM_012962716.2 | XP_012818170.1 | 4 - 5 | 737 | 73 | ||
| XM_012962717.2 | XP_012818171.1 | 3 - 4 | 711 | 73 | ||
| XM_012962718.2 | XP_012818172.1 | 3 - 4 | 711 | 73 | ||
| GenBank mRNA | BC159156.1 | - | 3 - 4 | 652 | 73 | |
Set Membership: |
|