Search
Search

uck2| Gene Symbol: uck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126769.1 | NP_001120241.1 | 1 - 2 | 238 | 79 | |
| GenBank mRNA | BC160433.1 | - | 1 - 2 | 238 | 79 | |
| BC161596.1 | - | 1 - 2 | 188 | 79 | ||
| CT025214.2 | - | 1 - 2 | 121 | 79 | ||
Set Membership: |
|