Search
Search

plppr3| Gene Symbol: plppr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126788.1 | NP_001120260.1 | 5 - 6 | 676 | 60 | |
| XM_018090851.1 | XP_017946340.1 | 5 - 6 | 699 | 60 | ||
| GenBank mRNA | BC160501.1 | - | 5 - 6 | 676 | 60 | |
Set Membership: |
|