Search
Search

mettl17| Gene Symbol: mettl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127148.1 | NP_001120620.1 | 6 - 7 | 658 | 65 | |
| GenBank mRNA | BC161764.1 | - | 6 - 7 | 658 | 65 | |
| CU075325.1 | - | 6 - 7 | 662 | 65 | ||
Set Membership: |
|