Search
Search

ptprn| Gene Symbol: ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127959.1 | NP_001121431.1 | 12 - 13 | 1841 | 71 | |
| GenBank mRNA | BC166171.1 | - | 12 - 13 | 1841 | 71 | |
Set Membership: |
|