Search
Search

zer1| Gene Symbol: zer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127966.1 | NP_001121438.1 | 4 - 5 | 897 | 61 | |
| XM_012968053.1 | XP_012823507.1 | 5 - 6 | 1078 | 61 | ||
| XM_012968054.1 | XP_012823508.1 | 5 - 6 | 1077 | 61 | ||
| XM_012968055.2 | XP_012823509.1 | 5 - 6 | 1137 | 61 | ||
| XM_012968056.2 | XP_012823510.1 | 5 - 6 | 1082 | 61 | ||
| XM_012968057.2 | XP_012823511.1 | 4 - 5 | 973 | 61 | ||
| GenBank mRNA | BC166186.1 | - | 4 - 5 | 897 | 61 | |
Set Membership: |
|