Search
Search

nme9| Gene Symbol: nme9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127984.1 | NP_001121456.1 | 7 - 8 | 640 | 75 | |
| XM_012970127.2 | XP_012825581.1 | 7 - 8 | 668 | 75 | ||
| GenBank mRNA | BC166216.1 | - | 7 - 8 | 640 | 75 | |
Set Membership: |
|