Search
Search

dennd5a| Gene Symbol: dennd5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127996.1 | NP_001121468.1 | 5 - 6 | 1244 | 69 | |
| XM_012960650.2 | XP_012816104.2 | 5 - 6 | 1599 | 69 | ||
| XM_018090113.1 | XP_017945602.1 | 5 - 6 | 1101 | 69 | ||
| XM_018090114.1 | XP_017945603.1 | 4 - 5 | 1255 | 69 | ||
| XM_018090115.1 | XP_017945604.1 | 4 - 5 | 1255 | 69 | ||
| XM_018090117.1 | XP_017945606.1 | 7 - 8 | 1297 | 69 | ||
| GenBank mRNA | BC166235.1 | - | 5 - 6 | 1244 | 69 | |
Set Membership: |
|