Search
Search

ak3| Gene Symbol: ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130221.1 | NP_001123693.1 | 2 - 3 | 326 | 104 | |
| XM_012956966.2 | XP_012812420.1 | 2 - 3 | 104 | |||
| GenBank mRNA | BC167275.1 | - | 2 - 3 | 326 | 104 | |
Set Membership: |
|