Search
Search

epm2a| Gene Symbol: epm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130223.1 | NP_001123695.1 | 3 - 4 | 530 | 71 | |
| XM_012962766.2 | XP_012818220.1 | 3 - 4 | 513 | 71 | ||
| GenBank mRNA | BC167294.1 | - | 3 - 4 | 530 | 71 | |
| BC167295.1 | - | 3 - 4 | 530 | 71 | ||
Set Membership: |
|