Search
Search

nme6| Gene Symbol: nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130237.1 | NP_001123709.1 | 1 - 2 | 113 | 107 | |
| XM_012958647.2 | XP_012814101.1 | 2 - 3 | 302 | 107 | ||
| XM_018091950.1 | XP_017947439.1 | 2 - 3 | 125 | 107 | ||
| GenBank mRNA | BC167315.1 | - | 1 - 2 | 113 | 107 | |
Set Membership: |
|