Search
Search

hbs1l| Gene Symbol: hbs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130349.1 | NP_001123821.1 | 1 - 2 | 136 | 69 | |
| XM_012962789.2 | XP_012818243.1 | 1 - 2 | 224 | 69 | ||
| XM_012962790.2 | XP_012818244.1 | 1 - 2 | 224 | 69 | ||
| GenBank mRNA | BC158221.1 | - | 1 - 2 | 183 | 69 | |
| BC167628.1 | - | 1 - 2 | 136 | 69 | ||
| CR942563.2 | - | 1 - 2 | 108 | 69 | ||
| CU075830.1 | - | 1 - 2 | 121 | 69 | ||
Set Membership: |
|