Search
Search

rptor| Gene Symbol: rptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130371.1 | NP_001123843.1 | 23 - 24 | 2924 | 95 | |
| XM_012971553.2 | XP_012827007.1 | 25 - 26 | 3153 | 95 | ||
| XM_012971554.2 | XP_012827008.1 | 25 - 26 | 3144 | 95 | ||
| XM_012971555.2 | XP_012827009.1 | 25 - 26 | 3153 | 95 | ||
| XM_012971556.2 | XP_012827010.1 | 25 - 26 | 3144 | 95 | ||
| XM_012971557.2 | XP_012827011.1 | 24 - 25 | 3006 | 95 | ||
| GenBank mRNA | BC167708.1 | - | 23 - 24 | 2924 | 95 | |
Set Membership: |
|