Search
Search

frs2| Gene Symbol: frs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134809.1 | NP_001128281.1 | 5 - 6 | 584 | 94 | |
| XM_012958358.2 | XP_012813812.1 | 5 - 6 | 743 | 94 | ||
| GenBank mRNA | BC168573.1 | - | 5 - 6 | 584 | 94 | |
| CT030446.1 | - | 5 - 6 | 607 | 94 | ||
Set Membership: |
|