Search
Search

cdk12| Gene Symbol: cdk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134811.1 | NP_001128283.1 | 4 - 5 | 2386 | 107 | |
| XM_012971492.1 | XP_012826946.1 | 4 - 5 | 2194 | 107 | ||
| GenBank mRNA | BC168577.1 | - | 4 - 5 | 2386 | 107 | |
| CT025435.2 | - | 4 - 5 | 1333 | 107 | ||
Set Membership: |
|