Search
Search

epha4| Gene Symbol: epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142103.1 | NP_001135575.1 | 16 - 17 | 2914 | 89 | |
| XM_012970177.1 | XP_012825631.1 | 16 - 17 | 2954 | 89 | ||
| XM_012970179.2 | XP_012825633.1 | 16 - 17 | 2952 | 89 | ||
| XM_018093809.1 | XP_017949298.1 | 16 - 17 | 2987 | 89 | ||
| GenBank mRNA | BC168080.1 | - | 16 - 17 | 2914 | 89 | |
Set Membership: |
|