Search
Search

cdh17| Gene Symbol: cdh17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142108.1 | NP_001135580.1 | 12 - 13 | 1628 | 92 | |
| GenBank mRNA | BC168087.1 | - | 12 - 13 | 1628 | 92 | |
Set Membership: |
|