Search
Search

bag5| Gene Symbol: bag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131044.1 | NP_001124516.1 | 1 | 1046 | 96 | |
| XM_012967917.1 | XP_012823371.1 | 2 | 1200 | 96 | ||
| GenBank mRNA | BC167949.1 | - | 1 | 1046 | 96 | |
| CU025116.1 | - | 2 | 1205 | 96 | ||
Set Membership: |
|