Search
Search

sid1| Gene Symbol: sid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146067.1 | NP_001139539.1 | 721 | 78 | ||
| GenBank mRNA | BT063357.1 | - | 721 | 78 | ||
| EU974186.1 | - | 676 | 78 | |||
| FJ686697.1 | - | 676 | 78 | |||
Set Membership: |
|