Search
Search

LOC100283820| Gene Symbol: LOC100283820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156718.1 | NP_001150190.1 | 887 | 73 | ||
| GenBank mRNA | BT065050.1 | - | 1048 | 73 | ||
| EU966063.1 | - | 887 | 73 | |||
Set Membership: |
|