Search
Search

LOC100191409| Gene Symbol: LOC100191409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136843.1 | NP_001130315.1 | 278 | 112 | ||
| GenBank mRNA | BT033491.1 | - | 278 | 112 | ||
| BT063725.1 | - | 177 | 112 | |||
| BT067460.1 | - | 225 | 112 | |||
| BT067481.1 | - | 210 | 112 | |||