Search
Search

cad1| Gene Symbol: cad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137309.1 | NP_001130781.1 | 210 | 77 | ||
| GenBank mRNA | BT034128.1 | - | 210 | 77 | ||
| BT040774.1 | - | 207 | 77 | |||
Set Membership: |
|