Search
Search

sad1| Gene Symbol: sad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137318.1 | NP_001130790.1 | 1146 | 74 | ||
| GenBank mRNA | BT034140.1 | - | 1146 | 74 | ||
| BT036418.1 | - | 945 | 74 | |||
Set Membership: |
|