Search
Search

LOC100276550| Gene Symbol: LOC100276550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001150312.1 | NP_001143784.1 | 962 | 96 | ||
| GenBank mRNA | EU963179.1 | - | 962 | 96 | ||
| EU963759.1 | - | 907 | 96 | |||