Search
Search

magi42672| Gene Symbol: magi42672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155829.1 | NP_001149301.1 | 249 | 94 | ||
| GenBank mRNA | EU952553.1 | - | 225 | 94 | ||
| EU962806.1 | - | 249 | 94 | |||
Set Membership: |
|