Search
Search

LOC100285233| Gene Symbol: LOC100285233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158127.1 | NP_001151599.1 | 1276 | 103 | ||
| GenBank mRNA | EU971367.1 | - | 1276 | 103 | ||
Set Membership: |
|