Search
Search

LOC100284450| Gene Symbol: LOC100284450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157345.1 | NP_001150817.1 | 618 | 92 | ||
| GenBank mRNA | BT042398.1 | - | 576 | 92 | ||
| EU968434.1 | - | 618 | 92 | |||
Set Membership: |
|