Search
Search

LOC100280991| Gene Symbol: LOC100280991 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153911.1 | NP_001147383.1 | 488 | 95 | ||
| GenBank mRNA | BT061636.1 | - | 445 | 95 | ||
| EU955101.1 | - | 488 | 95 | |||
Set Membership: |
|