Search
Search

qsox1| Gene Symbol: qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112299.1 | NP_001105769.1 | 756 | 83 | ||
| GenBank mRNA | AY739305.1 | - | 756 | 83 | ||
| BT063166.1 | - | 683 | 83 | |||
Set Membership: |
|