Search
Search

pco121171| Gene Symbol: pco121171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158891.1 | NP_001152363.1 | 1555 | 98 | ||
| GenBank mRNA | BT066332.1 | - | 1476 | 98 | ||
| EU975110.1 | - | 1555 | 98 | |||
Set Membership: |
|