Search
Search

LOC100280414| Gene Symbol: LOC100280414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153337.1 | NP_001146809.1 | 737 | 79 | ||
| GenBank mRNA | BT056188.1 | - | 737 | 79 | ||
| BT060890.1 | - | 81 | 79 | |||
Set Membership: |
|