Search
Search

LOC100285136| Gene Symbol: LOC100285136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158030.1 | NP_001151502.1 | 1190 | 112 | ||
| GenBank mRNA | EU970985.1 | - | 1190 | 112 | ||
Set Membership: |
|