Search
Search

LOC100276194| Gene Symbol: LOC100276194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001150039.1 | NP_001143511.1 | 1233 | 130 | ||
| GenBank mRNA | BT067810.1 | - | 1213 | 130 | ||
| EU960582.1 | - | 1233 | 130 | |||
Set Membership: |
|