Search
Search

igl1| Gene Symbol: igl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111532.1 | NP_001105002.1 | 674 | 57 | ||
| GenBank mRNA | AF271384.1 | - | 674 | 57 | ||
| AY103922.1 | - | 667 | 57 | |||
Set Membership: |
|