Search
Search

serk1| Gene Symbol: serk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111662.1 | NP_001105132.1 | 443 | 68 | ||
| GenBank mRNA | AJ277702.1 | - | 443 | 68 | ||
| BT042243.1 | - | 687 | 68 | |||
Set Membership: |
|