Search
Search

gbp2| Gene Symbol: gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111895.1 | NP_001105365.1 | 1189 | 87 | ||
| GenBank mRNA | AY109568.1 | - | 1189 | 87 | ||
| U12233.1 | - | 1189 | 87 | |||
Set Membership: |
|