Search
Search

dek1| Gene Symbol: dek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112058.1 | NP_001105528.1 | 3454 | 73 | ||
| GenBank mRNA | AY061805.1 | - | 3454 | 73 | ||
| AY061806.1 | - | 3454 | 73 | |||
Set Membership: |
|