Search
Search

mlo1| Gene Symbol: mlo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112190.1 | NP_001105660.1 | 1188 | 65 | ||
| GenBank mRNA | AY029312.2 | - | 1188 | 65 | ||
| AY110401.1 | - | 1160 | 65 | |||
| BT036101.1 | - | 1293 | 65 | |||
Set Membership: |
|