Search
Search

csu842| Gene Symbol: csu842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138278.1 | NP_001131750.1 | 546 | 71 | ||
| GenBank mRNA | BT034536.1 | - | 521 | 71 | ||
| BT035292.1 | - | 546 | 71 | |||
| BT041076.1 | - | 546 | 71 | |||
| EU953495.1 | - | 564 | 71 | |||
Set Membership: |
|