Search
Search

LOC100217038| Gene Symbol: LOC100217038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143410.1 | NP_001136882.1 | 1543 | 89 | ||
| GenBank mRNA | BT037814.1 | - | 1543 | 89 | ||
| EU953282.1 | - | 1543 | 89 | |||