Search
Search

LOC100283240| Gene Symbol: LOC100283240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156142.1 | NP_001149614.1 | 1148 | 110 | ||
| GenBank mRNA | EU963972.1 | - | 1148 | 110 | ||
| EU972736.1 | - | 1191 | 110 | |||
Set Membership: |
|