Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01118845_m1
          See other CENPF GE Assays ›
          Gene Symbol:
          CENPF
          Gene Name:
          centromere protein F
          Gene Aliases:
          CENF, CILD31, PRO1779, STROMS, hcp-1
          Chromosome Location:
          Chr.1: 214603195 - 214664571 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01118845_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CENPF
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016343.4 NP_057427.3 5 - 6 742 77
          XM_011509082.4 XP_011507384.1 5 - 6 742 77
          XM_017000086.3 XP_016855575.1 5 - 6 4175 77
          GenBank mRNA AK307690.1 - 5 - 6 731 77
          U19769.1 - 744 77
          U30872.1 - 5 - 6 646 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100011581.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001791.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CENPF

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          kidney development
          chromosome segregation
          mitotic spindle assembly checkpoint
          muscle organ development
          response to xenobiotic stimulus
          regulation of G2/M transition of mitotic cell cycle
          protein transport
          regulation of striated muscle tissue development
          ventricular system development
          cell differentiation
          negative regulation of transcription, DNA-templated
          cell division
          metaphase plate congression
          kinetochore assembly
          DNA biosynthetic process
          chromatin binding
          protein binding
          microtubule binding
          protein homodimerization activity
          dynein complex binding
          DNA-binding transcription factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000467765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000467765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000467765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000467765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000469862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000469862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000469862.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000495259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000495259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000495259.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000614578.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000614578.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 ENST00000614578.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01028005.hg.1 ENST00000366955.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01028005.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01028005.hg.1 TC01001791.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01028004.hg.1 ENST00000366955.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01028004.hg.1 NM_016343.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01028004.hg.1 TC01001791.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100185276.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100185276.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100185276.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100185276.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100185276.hg.1 TC0100011581.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000464322.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000467765.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000467765.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000467765.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000467765.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000469862.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000469862.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000469862.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000495259.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000495259.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000495259.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000614578.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000614578.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 ENST00000614578.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay TC0100011581.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100185277.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100185277.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100185277.hg.1 ENST00000366955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100185277.hg.1 NM_016343.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100185277.hg.1 TC0100011581.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline