Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04176264_s1
          See other P2RY2 GE Assays ›
          Gene Symbol:
          P2RY2
          Gene Name:
          purinergic receptor P2Y2
          Gene Aliases:
          HP2U, P2RU1, P2U, P2U1, P2UR, P2Y2, P2Y2R
          Chromosome Location:
          Chr.11: 73200416 - 73246743 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04176264_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2RY2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002564.3 NP_002555.3 3 1137 82
          NM_176071.2 NP_788085.2 3 1202 82
          NM_176072.2 NP_788086.2 3 1271 82
          XM_005274019.4 XP_005274076.1 3 1089 82
          XM_005274020.4 XP_005274077.1 3 2676 82
          XM_005274021.4 XP_005274078.1 3 2270 82
          XM_011545074.2 XP_011543376.1 3 1024 82
          XM_017017839.1 XP_016873328.1 4 1149 82
          GenBank mRNA AK313942.1 - 3 1098 82
          AY136753.1 - 1 803 82
          BC012104.1 - 3 1270 4561430 82
          BC028135.1 - 3 1202 5215817 82
          FJ599699.1 - 1 254 82
          FJ599700.1 - 1 257 82
          U07225.1 - 3 1048 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100008378.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000772.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2RY2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          cellular ion homeostasis
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          aging
          positive regulation of neuron projection development
          positive regulation of smooth muscle cell migration
          sensory perception of pain
          negative regulation of ossification
          positive regulation of prostaglandin secretion
          positive regulation of chemokine production
          G-protein coupled purinergic nucleotide receptor signaling pathway
          wound healing
          regulation of vasodilation
          positive regulation of penile erection
          positive regulation of mucus secretion
          response to growth factor
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to purine-containing compound
          receptor activity
          ATP binding
          G-protein coupled purinergic nucleotide receptor activity
          UTP-activated nucleotide receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000565433.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000565433.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009925.hg.1 ENST00000311131.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009925.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009925.hg.1 ENST00000393597.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009925.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009925.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009925.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009925.hg.1 TC11000772.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161923.hg.1 TC1100008378.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000565433.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000565433.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline