Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441087_m1
          See other FOXO3 GE Assays ›
          Gene Symbol:
          Foxo3
          Gene Name:
          forkhead box O3
          Gene Aliases:
          Fkhrl1, Foxo3a
          Chromosome Location:
          Chr.20: 46428078 - 46519156 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441087_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Foxo3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106395.1 NP_001099865.1 3 - 4 2385 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000833.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ovulation from ovarian follicle
          initiation of primordial ovarian follicle growth
          antral ovarian follicle growth
          oocyte maturation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          regulation of translation
          DNA damage response, signal transduction by p53 class mediator
          cellular response to oxidative stress
          glucose homeostasis
          positive regulation of neuron apoptotic process
          positive regulation of erythrocyte differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of canonical Wnt signaling pathway
          neuronal stem cell population maintenance
          extrinsic apoptotic signaling pathway in absence of ligand
          regulation of neural precursor cell proliferation
          regulation of reactive oxygen species metabolic process
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter binding
          transcription cofactor binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein kinase binding
          chromatin DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P085072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360660 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86385 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108577 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66153 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085072 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P085072 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16998 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67555 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74740 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38138 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833817 - -
          Affymetrix Rat Gene 1.0 ST Array 10833815 - -
          Affymetrix Rat Gene 1.0 ST Array 10833816 - -
          Affymetrix Rat Gene 1.1 ST Array 10833814 - -
          Rat Genome 230 2.0 Array 1376210_PM_at - -
          Rat Genome 230 2.0 Array 1376229_at - -
          Rat Genome 230 2.0 Array 1376229_PM_at - -
          Rat Genome 230 2.0 Array 1376210_at - -
          Affymetrix HT Rat Focus Array 1376210_PM_at - -
          Affymetrix HT Rat Focus Array 1376229_PM_at - -
          Affymetrix HT Rat Focus Array 1376210_at - -
          Affymetrix HT Rat Focus Array 1376229_at - -
          Affymetrix Rat Clariom S Assay PSR2000007027.rn.1 TC2000000833.rn.1 RTA TC
          Rat Expression Set 230 1376210_PM_at - -
          Rat Expression Set 230 1376210_at - -
          Rat Expression Set 230 1376229_PM_at - -
          Rat Expression Set 230 1376229_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833814 - -
          Rat Genome U34 Set rc_AI231684_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21576465 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374077 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline